Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556081 Supplier Invitrogen™ Supplier No. PA556081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLKSHSRDRN GLKKSNSPVH HNILAPVPGP APAHQRAVQN LQQHNLIVHF QANEDTPKSV PEKNLFKEAC EKRAQDLEMM ADDNIEDSTA RLDTQHSEDM NATRSEEQFH Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 72%.

This gene encodes a member of the kelch family of proteins, which are characterized by kelch repeat motifs and a POZ/BTB protein-binding domain. It is thought that kelch repeats are actin binding domains. However, the specific function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Klhl4
Gene Accession No. Q9C0H6
Gene Alias B130014A11; C130018J01Rik; DKELCHL; kelch like family member 4; kelch-like 4; kelch-like family member 4; kelch-like protein 4; KHL4; KIAA1687; KLHL4; RGD1565441
Gene Symbols Klhl4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHL4. Recombinant protein control fragment (Product #RP-91932).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56062
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.