Learn More
Invitrogen™ KLHL41 Monoclonal Antibody (1H3)

Description
Peptide Sequence: RTDKENRVKN LSEVFDCIRF RLMTEKYFKD HVEKDDIIKS NPDLQKKIKV LKDAFAGKLP EPSKNAAKTG AGEVNGDVGD EDLLPGYLND IPRHGMFVKD.
Specifications
Specifications
| Antigen | KLHL41 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1H3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | KLHL41 |
| Gene Accession No. | O60662 |
| Gene Alias | Gm112; KBTBD10; kelch like family member 41; kelch related protein 1; kelch repeat and BTB (POZ) domain containing 10; kelch repeat and BTB domain-containing protein 10; kelch-like 41; kelch-like family member 41; kelch-like protein 41; Kelch-related protein 1; Kel-like protein 23; Klhl41; KRP1; sarcomeric muscle protein; Sarcosin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.