Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554339 Supplier Invitrogen™ Supplier No. PA554339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAGEVNGDVG DEDLLPGYLN DIPRHGMFVK DLILLVNDTA AVAYDPTENE CYLTALAEQI PRNHSSIVTQ QNQI Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL41
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL41
Gene Accession No. O60662
Gene Alias Gm112; KBTBD10; kelch like family member 41; kelch related protein 1; kelch repeat and BTB (POZ) domain containing 10; kelch repeat and BTB domain-containing protein 10; kelch-like 41; kelch-like family member 41; kelch-like protein 41; Kelch-related protein 1; Kel-like protein 23; Klhl41; KRP1; sarcomeric muscle protein; Sarcosin
Gene Symbols KLHL41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHL41. Recombinant protein control fragment (Product #RP-93587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10324
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.