Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHL41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554498
Description
Immunogen sequence: GDKSLPCHRL ILSACSPYFR EYFLSEIDEA KKKEVVLDNV DPAILDLIIK YLYSASIDLN DGNVQD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.
Specifications
KLHL41 | |
Polyclonal | |
Unconjugated | |
KLHL41 | |
Gm112; KBTBD10; kelch like family member 41; kelch related protein 1; kelch repeat and BTB (POZ) domain containing 10; kelch repeat and BTB domain-containing protein 10; kelch-like 41; kelch-like family member 41; kelch-like protein 41; Kelch-related protein 1; Kel-like protein 23; Klhl41; KRP1; sarcomeric muscle protein; Sarcosin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60662 | |
KLHL41 | |
Recombinant protein corresponding to Human KLHL41. Recombinant protein control fragment (Product #RP-93585). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction