Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHL41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540849
Description
Peptide sequence: RFRLMTEKYF KDHVEKDDII KSNPDLQKKI KVLKDAFAGK LPEPSKNAAK Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.
Specifications
KLHL41 | |
Polyclonal | |
Unconjugated | |
KLHL41 | |
Gm112; KBTBD10; kelch like family member 41; kelch related protein 1; kelch repeat and BTB (POZ) domain containing 10; kelch repeat and BTB domain-containing protein 10; kelch-like 41; kelch-like family member 41; kelch-like protein 41; Kelch-related protein 1; Kel-like protein 23; Klhl41; KRP1; sarcomeric muscle protein; Sarcosin | |
Rabbit | |
Affinity chromatography | |
RUO | |
10324 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60662 | |
KLHL41 | |
Synthetic peptide directed towards the middle region of human KLHL41. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction