Learn More
KLHL42 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305649AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.Specifications
KLHL42 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8BFQ9, Q9P2K6 | |
Klhl42 | |
Between 400 and 450 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
KLHL42 | |
Ctb9, kelch domain-containing protein 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
232539, 57542 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.