Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556168 Supplier Invitrogen™ Supplier No. PA556168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRVTHLLNQD TLTVRAEDQV YDAAVRWLKY DEPNRQPFMV DILAKVRFPL ISKNFLSKTV QAEPLIQDNP ECLKMVISGM RYHLLSPEDR EELVDGTRPR RKKHDYRIAL FGGSQPQSCR YFNPKDYSWT DI Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Defects in KLHL7 are the cause of retinitis pigmentosa type 42 (RP42). The specific function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL7
Gene Accession No. Q8IXQ5
Gene Alias 2700038B03Rik; BTB/POZ domain containing protein; D5Ertd363e; kelch like family member 7; kelch/BTB; kelch-like 6; kelch-like 7; kelch-like family member 7; kelch-like protein 7; KLHL6; Klhl7; LOW QUALITY PROTEIN: kelch-like protein 7; SBBI26
Gene Symbols KLHL7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHL7. Recombinant protein control fragment (Product #RP-92693).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.