Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL7 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | KLHL7 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
KLHL7 Polyclonal specifically detects KLHL7 in Human, Mouse samples. It is validated for Western Blot.Specifications
KLHL7 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
kelch (Drosophila)-like 6, kelch/BTB, kelch-like 6, kelch-like 7 (Drosophila), kelch-like protein 7, KLHL6, RP42, SBBI26 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human KLHL7 (NP_061334.4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
55975 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title