Learn More
Invitrogen™ KLHL8 Polyclonal Antibody

Description
Immunogen sequence: HFHPSNCLAV RAFAESHNRI DLMDMADQYA CDHFTEVVEC EDFVSVSPQH LHKLLSSSDL NIENEKQVYN AAIKWLLANP QHHSKWLDET LAQVRLPLLP VDFLMGVVAK EQIVKQNLKC RDLLDEARNY HL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Specifications
Specifications
| Antigen | KLHL8 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KLHL8 |
| Gene Accession No. | Q9P2G9 |
| Gene Alias | 2310001P09Rik; D5Ertd431e; kelch like family member 8; kelch-like 8; kelch-like family member 8; kelch-like protein 8; KIAA1378; klhl8; zgc:158266 |
| Gene Symbols | KLHL8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.