Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569013 Supplier Invitrogen™ Supplier No. PA569013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%; Zebrafish: 77%.

Kelch-like protein 8 is a protein that in humans is encoded by the KLHL8 gene. The mRNA contains a repetitive element in its 3-prime UTR, and the deduced 451-amino acid protein shares 36% identity with the actin-binding protein MAYVEN. RT-PCR ELISA detected highest expression in adult brain, liver, skeletal muscle, kidney, and ovary, and in several specific brain regions, predominantly amygdala and thalamus. Moderate expression was detected in all other adult and fetal tissues examined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KLHL8
Gene Accession No. Q9P2G9
Gene Alias 2310001P09Rik; D5Ertd431e; kelch like family member 8; kelch-like 8; kelch-like family member 8; kelch-like protein 8; KIAA1378; klhl8; zgc:158266
Gene Symbols KLHL8
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KLHL8.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57563
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.