Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590126 Supplier Thermo Scientific Supplier No. PA590126
Add to Cart
Add to Cart

KLHL8 Polyclonal Antibody

KLHL8 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KLHL8
Gene Accession No. P59280, Q9P2G9
Gene Alias 2310001P09Rik; D5Ertd431e; kelch like family member 8; kelch-like 8; kelch-like family member 8; kelch-like protein 8; KIAA1378; klhl8; zgc:158266
Gene Symbols KLHL8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 271-620 of human KLHL8 (NP_0658543).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246293, 289457, 57563
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.