Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK3 Rabbit anti-Human, Unconjugated, Clone: RM323, Abnova™

Catalog No. 89339196 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-196 Supplier Abnova Corporation Supplier No. MAB21979
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

KLK3 Monoclonal antibody specifically detects KLK3 in Human samples. It is validated for WB, ELISA, IHC-P.

Specifications

Antigen KLK3
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM323
Conjugate Unconjugated
Description kallikrein-related peptidase 3
Gene KLK3
Gene Alias APS, KLK2A1, PSA, hK3
Host Species Rabbit
Immunogen Native PSA protein purified from human seminal fluid
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 354
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.