Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK8 Mouse anti-Human, Clone: OTI3C10, liquid, TrueMAB™
SDP

Catalog No. 501677272
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7272 Supplier OriGene Supplier No. TA600206
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.

Specifications

Antigen KLK8
Applications ELISA
Classification Monoclonal
Clone OTI3C10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <5% trehalose and 0.05% sodium azide
Gene KLK8
Gene Accession No. O60259
Gene Alias HNP, NP, NRPN, PRSS19, TADG14
Gene Symbols KLK8
Host Species Mouse
Immunogen Full length human recombit protein of human KLK8 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11202
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.