Learn More
KLK8 Mouse anti-Human, Clone: OTI5D1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801597

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.Specifications
KLK8 | |
Monoclonal | |
Unconjugated | |
KLK8 | |
HNP, NP, NRPN, PRSS19, TADG14 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11202 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5D1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60259 | |
KLK8 | |
Human recombit protein fragment corresponding to amino acids 33-260 of human KLK8 produced in SF9 cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.