Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986871
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6871 Supplier Bioss Supplier No. BS5871R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.

Specifications

Antigen KLK8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KLK8
Gene Accession No. O60259
Gene Alias Brain Serine protease 1; bsp1; H NRPN; hK8; HNP; kallikrein 8; kallikrein 8 (neuropsin/ovasin); kallikrein related peptidase 8; kallikrein related-peptidase 8; Kallikrein8; Kallikrein-8; kallikrein-related peptidase 8; Klk8; mK8; Neuropsin; NP; NRPN; Ovasin; PRO322; protease, serine, 19 (neuropsin); Prss19; Serine protease 19; Serine protease TADG-14; TADG14; Tumor-associated differentially expressed gene 14 protein; UNQ283; UNQ283/PRO322
Gene Symbols KLK8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse KLK8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11202
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.