Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK9 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278161
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8161 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8161 Supplier Leinco Technologies Inc Supplier No. 56239100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLK9 Polyclonal antibody specifically detects KLK9 in Human samples. It is validated for Western Blot

Specifications

Antigen KLK9
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene KLK9
Gene Accession No. Q9UKQ9
Gene Alias 1200016C12Rik; AI324041; kallikrein 9; kallikrein related peptidase 9; kallikrein related-peptidase 9; kallikrein-9; Kallikrein-like protein 3; kallikrein-related peptidase 9; KLK9; KLK-9; KLKL3; KLK-L3
Host Species Rabbit
Immunogen Peptide corresponding to aa 239-250 of human KLK9 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284366
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.