Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLLN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546688 Supplier Invitrogen™ Supplier No. PA546688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DSSGGKSSSS FARGALAWCR QRNPNPSCAA AETGARTSLP KERCRGWRLG Sequence homology: Human: 100%.

The protein encoded by this intronless gene is found in the nucleus, where it can inhibit DNA synthesis and promote S phase arrest coupled to apoptosis. The expression of this DNA binding protein is upregulated by transcription factor p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLLN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KLLN
Gene Accession No. B2CW77
Gene Alias CWS4; KILLIN; killin, p53-regulated DNA replication inhibitor; KLLN
Gene Symbols KLLN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KLLN.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100144748
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.