Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Klotho beta Antibody (NGM313) [Janelia Fluor™ 585], Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP328017JF585
Description
Klotho beta Monoclonal antibody specifically detects Klotho beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Klotho beta | |
| Monoclonal | |
| Janelia Fluor 585 | |
| beta-klotho, betaKlotho, BKL, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| Klotho Beta | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| NGM313 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 152831 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction