Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Klotho Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
$243.00 - $522.50
Specifications
Antigen | Klotho |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543727
|
Invitrogen™
MA543727 |
20 μL |
Each for $243.00
|
|
|||||
PIMA543726
|
Invitrogen™
MA543726 |
100 μL |
Each for $522.50
|
|
|||||
Description
KLOTHO is the systemic anti-aging hormone within the glycosidase1 superfamily. It encodes a type I membrane protein that is abundant in the kidney and brain. In mice, a deficiency in KLOTHO expression leads to various systemic phenotypes resembling human aging such as arteriosclerosis, osteoporosis, and skin atrophy together with growth retardation, short life-span and infertility. Transgenic mice overexpressing KLOTHO have an extended life span by inhibiting insulin/IGF1 signaling. KLOTHO is involved in the regulation of calcium/phosphorus homeostasis by inhibiting the synthesis of active vitamin D and identified as a potential tumor suppressor.Specifications
Klotho | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
alpha-kl; HFTC3; Kl; Klotho; Klotho peptide; secreted form of Klotho protein | |
KL | |
IgG1 κ | |
Protein A/G | |
Antibody |
C3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9UEF7 | |
9365 | |
Phe765-Ser981 (Accession Q9UEF7), with Two N-terminal Tags, His-tag and SUMO-tag. | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
KL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title