Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLRA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
| Antigen | KLRA1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
KLRA1 Polyclonal specifically detects KLRA1 in Human samples. It is validated for Western Blot.Specifications
| KLRA1 | |
| Polyclonal | |
| Rabbit | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| killer cell lectin-like receptor subfamily A pseudogene 1, KLRA#, KLRA1, Ly49, Ly-49L, LY49L, MGC126520, MGC126522 | |
| Synthetic peptides corresponding to KLRA1(killer cell lectin-like receptor subfamily A, member 1) The peptide sequence was selected from the N terminal of KLRA1. Peptide sequence NDQGEIYSTLRFLQSPSESQNRLRPDDTQRPGKTDDKEFSVPWHLIAVTL. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9UNG0 | |
| 10748 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title