Learn More
Invitrogen™ KLRA1 Polyclonal Antibody

Description
A suggested positive control is mouse spleen tissue lysate. PA5-20675 can be used with blocking peptide PEP-0793.
Specifications
Specifications
| Antigen | KLRA1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KLRA1P |
| Gene Accession No. | P20937 |
| Gene Alias | A1; CH29-493D4.3; class I MHC receptor KLRA22; killer cell lectin like receptor A1, pseudogene; killer cell lectin-like receptor 2; killer cell lectin-like receptor, subfamily A, member 1; killer cell lectin-like receptor, subfamily A, member 2; KLRA#; KLRA1; KLRA1P; Klra2; Klra22; KLRAP1; Ly49; Ly49 inhibitory receptor 8; Ly49a; ly-49a; Ly49i8; LY49L; Ly-49L; Ly49o>129>; Ly49v; lymphocyte antigen 49a; natural killer cell receptor Ly49A; novel killer cell lectin-like receptor subfamily A protein; t lymphocyte antigen A1; T-cell surface glycoprotein YE1/48 |
| Gene Symbols | Klra1, KLRA1P |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.