Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLRB1 Monoclonal Antibody (10-78), FITC

Catalog No. PIMA517539
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517539 500 μg
PIMA517538 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517539 Supplier Invitrogen™ Supplier No. MA517539
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLRB1 Monoclonal Antibody for Flow, Western Blot

KLRB1 (Killer cell lectin like receptor subfamily B, member 1, CD-161) protein is classified as a type II membrane protein because it has an external C terminus. KLRB1 cell surface antigen is expressed by almost all NK cells and in a small subset of CD3+ve T cells. KLRB1 is a homodimeric cell surface protein, comprising two chains with molecular weights ranging from 40-44kDa. KLRB1 plays an inhibitory role on natural killer (NK) cells. Activation of KLRB1 leads to acid sphingomyelnase/SMPD1 stimulation and an increase in intracellular ceramide. Moreover, there is also an activation of AKT1/PKB and RPS6KA1/RSK1 kinase stimulation, and T cell proliferation by anti-CD3. KLRB1 acts as a lectin that binds to the terminal carbohydrate Gal-alpha(1,3)Gal epitope and to the N-acetyllactosamine epitope. KLRB1 also binds to CLEC2D/LLT1 as a ligand, and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLRB1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 10-78
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Klrb1
Gene Accession No. A4KWA1, P27471, Q0ZUP0
Gene Alias 4930431A04Rik; Antigen 3.2.3; CD161; CD161 antigen-like family member A; CD161 antigen-like family member B; CD161a; CD161b; CLEC5B; C-type lectin domain family 5 member B; EMBL:AAQ11375.1}; Gm4696; HNKR-P1a; Immunoreceptor NKR-P1E; immunoreceptor NKR-P1G; inhibitory receptor NKR-P1B; killer cell lectin like receptor B1; killer cell lectin-like receptor subfamily A member 1 complex; killer cell lectin-like receptor subfamily B member 1; killer cell lectin-like receptor subfamily B member 1 complex; killer cell lectin-like receptor subfamily B member 1A; killer cell lectin-like receptor subfamily B member 1B; killer cell lectin-like receptor subfamily B member 1B allele A; Killer cell lectin-like receptor subfamily B member 1B allele B; killer cell lectin-like receptor subfamily B member 1B allele C; killer cell lectin-like receptor subfamily B member 1D; killer cell lectin-like receptor subfamily B member 1F; killer cell lectin-like receptor subfamily B member 1G; killer cell lectin-like receptor subfamily B, member 1; killer cell lectin-like receptor subfamily B, member 1A; Klrb1; Klrb1a; Klrb1b; klrb1b {ECO:0000312; Klrb1d; Klrb1f; Klrb1g; Klrb6; lectin-like transmembrane protein Nkrp1g; Ly55; Ly-55; Ly55a; ly-55A; Ly55b; ly-55b; Ly55d; ly-55d; lymphocyte antigen 55; lymphocyte antigen 55 complex, locus A; lymphocyte antigen 55 complex, locus B; lymphocyte antigen 55 complex, locus D; lymphocyte antigen 55A; lymphocyte antigen 55b; lymphocyte antigen 55d; MGC138614; natural killer cell receptor; natural killer cell receptor protein NKR-P1A; natural killer cell receptor protein NKR-P1B; natural killer cell receptor-P1; natural killer cell surface protein NKR-P1B allele B6; Natural killer cell surface protein NKR-P1B allele RNK/SD/BN/F344; natural killer cell surface protein NKR-P1B allele SJL/BALB; natural killer cell surface protein NKR-P1B allele TO; natural killer cell surface protein NKR-P1G; Natural killer cell surface protein P1-2; natural killer cell surface protein P1-3.2.3; Natural killer cell surface protein P1A; natural killer lectin-like receptor 1E; natural killer lectin-like receptor protein 1 e; NKR; NKR-P1; NKR-P1 2; NKR-P1 3.2.3; NKR-P1 34; NKR-P1.7; NKRP12; NKRP1A; NKR-P1A; Nkrp1-a; Nkrp1b; Nkr-p1b; Nkrp1-b; Nkrp1d; NKR-P1D; Nkrp1e; Nkr-p1e; Nkrp-1e; Nkrp1g; NKR-P1G; RGD:2975}
Gene Symbols Klrb1, Klrb1a, Klrb1b
Host Species Mouse
Immunogen 60 kDa disulfide linked homodimer NKR-P1.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25192, 362443, 689817
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.