Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRB1 Mouse anti-Human, Clone:4C6A11, Abnova™

Catalog No. 89334329 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-329 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-329 Supplier Abnova Corporation Supplier No. MAB17997
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human KLRB1.

Natural killer (NK) cells are lymphocytes that mediate cytotoxicity and secrete cytokines after immune stimulation. Several genes of the C-type lectin superfamily, including the rodent NKRP1 family of glycoproteins, are expressed by NK cells and may be involved in the regulation of NK cell function. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain. The KLRB1 protein is classified as a type II membrane protein because it has an external C terminus. [provided by RefSeq]

Specifications

Antigen KLRB1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4C6A11
Conjugate Unconjugated
Description killer cell lectin-like receptor subfamily B, member 1
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene KLRB1
Gene Alias CD161, CLEC5B, MGC138614, NKR, NKR-P1, NKR-P1A, NKRP1A, hNKR-P1A
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 67-225 of human KLRB1.
Quantity 100 μg
Gene ID (Entrez) 3820
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.