Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRB1 Mouse anti-Human, FITC, Clone: HP-3G10, Abnova™

Catalog No. 89934414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-414 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-414 Supplier Abnova Corporation Supplier No. MAB12235
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against KLRB1.

Natural killer (NK) cells are lymphocytes that mediate cytotoxicity and secrete cytokines after immune stimulation. Several genes of the C-type lectin superfamily, including the rodent NKRP1 family of glycoproteins, are expressed by NK cells and may be involved in the regulation of NK cell function. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain. The KLRB1 protein is classified as a type II membrane protein because it has an external C terminus. [provided by RefSeq]

Specifications

Antigen KLRB1
Applications Flow Cytometry
Classification Monoclonal
Clone HP-3G10
Conjugate FITC
Description Mouse monoclonal antibody raised against KLRB1.
Dilution Flow Cytometry The optimal working dilution should be determined by the end user.
Formulation In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer).
Gene Alias CD161/CLEC5B/MGC138614/NKR/NKR-P1/NKR-P1A/NKRP1A/hNKR-P1A
Gene Symbols KLRB1
Host Species Mouse
Immunogen KLRB1
Purification Method Affinity Chromatography
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3820
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.