Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLRC1/2/3 Antibody (20d5), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP128100PE
Description
KLRC1/2/3 Monoclonal specifically detects KLRC1/2/3 in Mouse samples. It is validated for Flow Cytometry.Specifications
KLRC1/2/3 | |
Monoclonal | |
PE | |
CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse NKG2 (Mr approx. 43 kDa) | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
20d5 | |
Flow Cytometry | |
KLRC1 | |
CHO cells transfected with B6 allele of NKG2A gene | |
0.25 mL | |
Signal Transduction | |
3821 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction