Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLRC2/KLRC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599852 Supplier Invitrogen™ Supplier No. PA599852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KLRC2/4.

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. NK cells preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The group, designated KLRC (NKG2) are expressed primarily in natural killer (NK) cells and encodes a family of transmembrane proteins characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The KLRC (NKG2) gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed on NK cells. KLRC2 alternative splice variants have been described but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLRC2/KLRC4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KLRC2
Gene Accession No. O43908, P26717
Gene Alias CD159 antigen-like family member C; CD159c; killer cell lectin like receptor C2; killer cell lectin like receptor C4; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 4; KLRC2; KLRC4; natual killer cell group 2-F; NK cell receptor C; NK cell receptor F; NKG2C; NKG2-C; NKG2-C type II integral membrane protein; NKG2-C-activating NK receptor; Nkg2E; NKG2F; NKG2-F; NKG2-F type II integral membrane protein; NKG2-F-activating NK receptor; rNKG2C
Gene Symbols KLRC2, KLRC4
Host Species Rabbit
Immunogen A synthesized peptide derived from human KLRC2(Accession P26717), corresponding to amino acid residues M1-Q40.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16642, 29684, 3822, 8302
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.