Learn More
Invitrogen™ KLRC4 Polyclonal Antibody

Description
Recommended positive controls: 293T. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | KLRC4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | KLRC4 |
| Gene Accession No. | O43908 |
| Gene Alias | killer cell lectin like receptor C4; killer cell lectin-like receptor subfamily C, member 4; KLRC4; natual killer cell group 2-F; NK cell receptor F; NKG2F; NKG2-F; NKG2-F type II integral membrane protein; NKG2-F-activating NK receptor |
| Gene Symbols | KLRC4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.