Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Klre-1 Antibody (854929) [Janelia Fluor™ 646], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7099J
Description
Klre-1 Monoclonal antibody specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Klre-1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3824 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
854929 | |
Flow Cytometry, CyTOF-ready | |
CD94 | |
Chinese hamster ovary cell line, CHO derived recombinant mouse Klre-1, Lys94-Lys226, Accession # NP_705818 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction