Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Klre-1 Antibody (854929), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7099MFV450
Description
Klre-1 Monoclonal antibody specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Klre-1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 854929 | |
| Flow Cytometry, CyTOF-ready | |
| CD94 | |
| Chinese hamster ovary cell line, CHO derived recombinant mouse Klre-1, Lys94-Lys226, Accession # NP_705818 | |
| RUO | |
| 3824 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction