Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Klre-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547899
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547899 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547899 Supplier Invitrogen™ Supplier No. PA547899
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human NKG2D and recombinant mouse NKG2D is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Klre-1
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Klre1
Gene Accession No. Q8CJC7
Gene Alias CD94; CD94 antigen; killer cell lectin like receptor D1; killer cell lectin-like receptor family E member 1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; Killer cell lectin-like receptor subfamily E member 1; KLRD1; Klre1; Klre-1; KP43; natural killer cells antigen CD94; NK cell receptor; NKG2I
Gene Symbols Klre1
Host Species Sheep
Immunogen CHO-derived recombinant mouse Klre-1 Lys94-Lys226.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243655
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.