Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5139821 Supplier Invitrogen™ Supplier No. PA5139821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GSCSNATQYE DTGDLKVNNG TRRNISNKDL CASRSADQTV LCQSEWLKYQ GKCYWFSNEM KSWSDSYVYC LER Highest antigen sequence indentity to the following orthologs: Mouse - 32%, Rat - 27%.

KLRF1, an activating homodimeric C-type lectin-like receptor (CTLR), is expressed on nearly all natural killer (NK) cells and stimulates their cytoxicity and cytokine release (Kuttruff et al., 2009 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLRF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLRF1
Gene Accession No. Q9NZS2
Gene Alias Activating coreceptor NKp80; CLEC5C; CLEC5CMGC119908; C-type lectin domain family 5 member C; killer cell lectin like receptor F1; killer cell lectin-like receptor F1; killer cell lectin-like receptor subfamily F member 1; killer cell lectin-like receptor subfamily F, member 1; KLRF1; Lectin-like receptor F1; MGC119907; MGC119909; ML; natural killer cell receptor; NKp80
Gene Symbols KLRF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLRF1. Recombinant protein control fragment (Product #RP-108908).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51348
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.