Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLRF2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549096

Catalog No. PA549096


Only null left
Add to Cart

Description

Description

KLRF2 Polyclonal Antibody for Western Blot

C-type lectin-like receptor involved in natural killer cell mediated cytotoxicity and cytokine secretion in keratinocytes via its interaction with CLEC2A.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KLRF2
Polyclonal
Unconjugated
KLRF2
activating coreceptor NKp65; killer cell lectin like receptor F2; killer cell lectin-like receptor subfamily F member 2; killer cell lectin-like receptor subfamily F, member 2; KLRF2; Lectin-like receptor F2; NKp65
Rabbit
Antigen affinity chromatography, Protein A
RUO
100431172
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
D3W0D1
KLRF2
KLH conjugated synthetic peptide between 4-30 amino acids from the N-terminal region of human KLRF2.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.