Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLRG1 Antibody (2F1), Alexa Fluor™ 405, Novus Biologicals™

Golden Syrian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP128115AF405
Description
KLRG1 Monoclonal specifically detects KLRG1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
KLRG1 | |
Monoclonal | |
Alexa Fluor 405 | |
2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
Golden Syrian Hamster | |
Protein A or G purified | |
RUO | |
Primary | |
KLRG1 (Mr 30-38 kDa) | |
Store at 4C in the dark. | |
IgG2 |
Flow Cytometry | |
2F1 | |
Flow Cytometry | |
KLRG1 | |
IL-2-activated NK cells (A-LAK cells) from B6 (H-2b) mice | |
0.25 mL | |
Immunology, Innate Immunity, Signal Transduction | |
10219 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction