Learn More
KLRG1 Antibody (2F1), Biotin, Novus Biologicals™
SDP
Golden Syrian Hamster Monoclonal Antibody
Supplier:  Novus Biologicals NBP128117
Catalog No. NBP128117
Only null left
Add to Cart
Description
Description
KLRG1 Monoclonal specifically detects KLRG1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications
Specifications
KLRG1
Monoclonal
0.5mg/mL
Flow Cytometry 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500
KLRG1
IL-2-activated NK cells (A-LAK cells) from B6 (H-2b) mice
0.25 mg
Immunology, Innate Immunity, Signal Transduction
10219
Human, Mouse
Purified
Flow Cytometry, Immunoprecipitation
2F1
Biotin
2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600
Golden Syrian Hamster
Protein A or G purified
RUO
Primary
KLRG1 (Mr 30-38 kDa)
Store at 4C. Do not freeze.
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only