Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLRG1 Antibody (2F1) [DyLight 350], Novus Biologicals™

Hamster Monoclonal Antibody
Supplier: Novus Biologicals FAB69441D
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
KLRG1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
2F1 | |
Flow Cytometry, CyTOF-ready | |
2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
IL-2 activated NK cells from C57/BL6 mice, Accession # O88713 | |
0.1 mL | |
Immunology, Innate Immunity, Signal Transduction | |
10219 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction