Learn More
Description
Specifications
Specifications
| Antigen | KLRG1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2F1 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 786 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA; 2F1 Ag |
| Host Species | Hamster |
| Immunogen | A-LAK from C57BL/6 mice |
| Show More |
Safety and Handling
For Reseach Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.