Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRG2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989822
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9822 Supplier Bioss Supplier No. BS16787R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen KLRG2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene KLRG2
Gene Accession No. A4D1S0
Gene Alias CLEC15B; C-type lectin domain family 15 member B; C-type lectin domain family 15, member B; killer cell lectin like receptor G2; Killer cell lectin-like receptor subfamily G member 2; killer cell lectin-like receptor subfamily G, member 2; KLRG2
Gene Symbols KLRG2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KLRG2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 346689
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.