Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMO Mouse anti-Human, Mouse, Rat, Clone: 2D2F2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | KMO |
|---|---|
| Clone | 2D2F2 |
| Concentration | 0.62 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6196
![]()
|
Proteintech Group Inc
600291IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.Specifications
| KMO | |
| 0.62 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| KMO, Kynurenine 3 hydroxylase, Kynurenine 3 monooxygenase | |
| Kmo | |
| IgG1 | |
| Caprylic Acid Precipitation | |
| Antibody |
| 2D2F2 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| O15229, O88867, Q91WN4 | |
| 59113, 8564, 98256 | |
| KMO Fusion Protein Ag1109 | |
| Primary | |
| -20°C | |
| KMO |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title