Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KmoRabbit anti-Human, Mouse, Polyclonal, Rockland™
SDP

Catalog No. 501992220
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2220 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2220 Supplier Rockland Immunochemicals Supplier No. 600401J25
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific towards Kynurenine 3-monooxygenase (KMO). A BLAST analysis was used to suggest cross-reactivity with Mouse based on 100% sequence homology. Cross-reactivity with KMO from other sources has not been determined.

Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.

Specifications

Antigen Kmo
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 50% glycerol and no preservative; pH 7.2
Gene KMO
Gene Accession No. O15229, Q91WN4
Gene Alias AI046660; dJ317G22.1; HAMAP-Rule:MF_03018}; KMO; kmo {ECO:0000312; Kynurenine 3-hydroxylase; kynurenine 3-hydroxylase {ECO:0000255; Kynurenine 3-monooxygenase; kynurenine 3-monooxygenase (kynurenine 3-hydroxylase); kynurenine 3-monooxygenase {ECO:0000255; RGD:620610}
Gene Symbols KMO
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8564, 98256
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.