Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KMP-11 Monoclonal Antibody (L-157)
GREENER_CHOICE

Catalog No. PIMA183818
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183818 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183818 Supplier Invitrogen™ Supplier No. MA183818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone L-157 was originally derived against Leishmania donovani lipophosphoglycan, but found to recognize the tightly associated LPGAP. Reconstitute with 0.5 mL of distilled water. Prior to reconstitution, store at 4ºC. After reconstitution, store undiluted at -20ºC, avoiding freeze/thaw cycles. Mouse anti Kinetoplastid Membrane Protein 11 antibody, clone L-157 recognizes Kinetoplastid Membrane Protein-11 (KMP-11), previously known as the lipophosphoglycan-associated protein (LPGAP), a 92 amino acid approximately 11 kDa membrane protein.

Kinetoplastid Membrane Protein-11 (KMP-11), previously known as the lipophosphoglycan-associated protein (LPGAP), is an 11 kDa membrane protein. KMP-11 is found in a wide variety of Kinetoplastid parasites including African trypanosomes, Leishmania and Crithidia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMP-11
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone L-157
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with no preservative
Gene KMP-11
Gene Accession No. Q36736
Gene Symbols KMP-11
Host Species Mouse
Immunogen L. donovani lipophosphoglycan semi purified from epimastigotes.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13387682
Target Species Protozoa
Content And Storage 4°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.