Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMT1A/SUV39H1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP121367
Description
KMT1A/SUV39H1 Polyclonal specifically detects KMT1A/SUV39H1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
KMT1A/SUV39H1 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
H3-K9-HMTase 1, Histone H3-K9 methyltransferase 1, histone-lysine N-methyltransferase SUV39H1, histone-lysine N-methyltransferase, H3 lysine-9 specific 1, KMT1AEC 2.1.1.43, Lysine N-methyltransferase 1A, MG44, Position-effect variegation 3-9 homolog, Su(var)3-9 homolog 1, suppressor of variegation 3-9 (Drosophila) homolog 1, Suppressor of variegation 3-9 homolog 1, suppressor of variegation 3-9 homolog 1 (Drosophila), Suppressor of variegation 3-9 homolog 1, Suvar) 3-9 homolog, SUV39H | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, European domestic ferret, African elephant and Northern white-cheeked gibbon. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 5- 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:1500 | |
O43463 | |
SUV39H1 | |
A synthetic peptide made to an N-terminal portion of the human KMT1A/SUV39H1 protein (between residues 1-100) [UniProt O43463] | |
0.1 mL | |
Cell Cycle and Replication, Chromatin Research | |
6839 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction