Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMT1A/SUV39H1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | KMT1A/SUV39H1 |
---|---|
Applications | ChIP Assay, Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
KMT1A/SUV39H1 Polyclonal specifically detects KMT1A/SUV39H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KMT1A/SUV39H1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
H3-K9-HMTase 1, Histone H3-K9 methyltransferase 1, histone-lysine N-methyltransferase SUV39H1, histone-lysine N-methyltransferase, H3 lysine-9 specific 1, KMT1AEC 2.1.1.43, Lysine N-methyltransferase 1A, MG44, Position-effect variegation 3-9 homolog, Su(var)3-9 homolog 1, suppressor of variegation 3-9 (Drosophila) homolog 1, Suppressor of variegation 3-9 homolog 1, suppressor of variegation 3-9 homolog 1 (Drosophila), Suppressor of variegation 3-9 homolog 1, Suvar) 3-9 homolog, SUV39H | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human KMT1A/SUV39H1 (NP_003164.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
ChIP Assay, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Chromatin Research | |
PBS with 50% glycerol, pH7.3. | |
6839 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title