Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMT2A/MLL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB600256
Description
KMT2A/MLL Polyclonal specifically detects KMT2A/MLL in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunoprecipitation.Specifications
KMT2A/MLL | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
ALL1, ALL-1MLL/GAS7, CXXC7TET1-MLL, CXXC-type zinc finger protein 7, EC 2.1.1.43, HRXFLJ11783, HTRX, HTRX1MLL-AF4 der(11) fusion protein, KMT2ACDK6/MLL fusion protein, Lysine N-methyltransferase 2A, MLL/GAS7 fusion protein, MLL/GMPS fusion protein, MLL1, MLL1A, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog), myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila), Trithorax-like protein, TRX1histone-lysine N-methyltransferase MLL, Zinc finger protein HRX | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
NB600-256 is specific for human MLL-1 protein. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Primates, White-tufted-ear marmoset, Black-handed spider monkey, Brown woolly monkey, Crab-eating macaque and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, Flow Cytometry, Immunoprecipitation, ChIP Assay | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Simple Western 1:10, Flow Cytometry, Immunoprecipitation 2-5 ug/mg of lysate, Chromatin Immunoprecipitation (ChIP) 1-5 ug/ml | |
Q03164 | |
MLL | |
The immunogen recognized by this antibody maps to a region between residues 2725 and 2775 of human myeloid/lymphoid or mixed-lineage leukemia 1 using the number given in Swiss-Prot entry Q03164 (GeneID 4297). | |
100 μL | |
Transcription Factors and Regulators | |
4297 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction