Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMT2B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $400.50
Specifications
Antigen | KMT2B |
---|---|
Dilution | Western Blot 1:500, ELISA 1:200-1:2000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
KMT2B Polyclonal antibody specifically detects KMT2B in Mouse samples. It is validated for Western Blot, ELISASpecifications
KMT2B | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
Whole antiserum | |
EC 2.1.1.43, histone-lysine N-methyltransferase MLL4, HRX2, KIAA0304WW domain-binding protein 7, Lysine N-methyltransferase 2D, mixed lineage leukemia gene homolog 2, MLL2, myeloid/lymphoid or mixed-lineage leukemia 4, Myeloid/lymphoid or mixed-lineage leukemia protein 4, Trithorax homolog 2, trithorax homologue 2, TRX2, WBP7, WBP-7, WW domain binding protein 7 | |
Polyclonal antibody raised in rabbit against mouse KMT2B (Myeloid/lymphoid or mixed-lineage leukemia protein 2), using two KLH-conjugated synthetic peptides containing an amino acid sequence from the N-terminal and one peptide containing a sequence from the central part of the protein, respectively. | |
Whole Antiserum |
Western Blot 1:500, ELISA 1:200-1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Mouse | |
Q9UMN6 | |
9757 | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title