Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KMT2C Monoclonal Antibody (1686CT202.60.69)
GREENER_CHOICE

Catalog No. PIMA524750
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524750 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524750 Supplier Invitrogen™ Supplier No. MA524750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KMT2C Monoclonal Antibody for Western Blot

Histone methyltransferase. Methylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Central component of the MLL2/3 complex, a coactivator complex of nuclear receptors, involved in transcriptional coactivation. KMT2C/MLL3 may be a catalytic subunit of this complex. May be involved in leukemogenesis and developmental disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2C
Applications Western Blot
Classification Monoclonal
Clone 1686CT202.60.69
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene KMT2C
Gene Accession No. Q8NEZ4
Gene Alias 2.1.1.43; ALR-like protein; HALR; Histone-lysine N-methyltransferase 2C; histone-lysine N-methyltransferase MLL3; histone-lysine N-methyltransferase, H3 lysine-4 specific; homologous to ALR protein; KIAA1506; KMT2C; lysine (K)-specific methyltransferase 2C; lysine methyltransferase 2C; Lysine N-methyltransferase 2C; MLL3; Myeloid/lymphoid or mixed-lineage leukemia protein 3
Gene Symbols KMT2C
Host Species Mouse
Immunogen human KMT2C recombinant protein.
Purification Method Protein G, Size-exclusion - Dialysis
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58508
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.