Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KMT2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568419 Supplier Invitrogen™ Supplier No. PA568419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KMT2C Polyclonal Antibody for IHC (P)

Histone methyltransferase. Methylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Central component of the MLL2/3 complex, a coactivator complex of nuclear receptors, involved in transcriptional coactivation. KMT2C/MLL3 may be a catalytic subunit of this complex. May be involved in leukemogenesis and developmental disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KMT2C
Gene Accession No. Q8NEZ4
Gene Alias 2.1.1.43; ALR-like protein; HALR; Histone-lysine N-methyltransferase 2C; histone-lysine N-methyltransferase MLL3; histone-lysine N-methyltransferase, H3 lysine-4 specific; homologous to ALR protein; KIAA1506; KMT2C; lysine (K)-specific methyltransferase 2C; lysine methyltransferase 2C; Lysine N-methyltransferase 2C; MLL3; Myeloid/lymphoid or mixed-lineage leukemia protein 3
Gene Symbols KMT2C
Host Species Rabbit
Immunogen Synthesized peptide (SQYRKMKTEW KSN) derived from human KMT2C.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58508
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.