Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMT2D Antibody (CL5346), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | KMT2D |
---|---|
Clone | CL5346 |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
KMT2D Monoclonal specifically detects KMT2D in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
KMT2D | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
MLL2 | |
IgG1 | |
Protein A purified |
CL5346 | |
Monoclonal | |
Purified | |
Human | |
8085 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AQLADTLFSKGLGPWDPPDNLAETQKPEQSSLVPGHLDQVNGQVVPEASQLSIKQEPREEPCALGAQSVKREANGEPIGAPGTSNHLLLAGPRSEAGHLLLQKLLRAKNVQLSTGRGSEGLRAEINGHI | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title