Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KMT2D Monoclonal Antibody (CL5346)
GREENER_CHOICE

Catalog No. PIMA535029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535029 Supplier Invitrogen™ Supplier No. MA535029
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: AQLADTLFSK GLGPWDPPDN LAETQKPEQS SLVPGHLDQV NGQVVPEASQ LSIKQEPREE PCALGAQSVK REANGEPIGA PGTSNHLLLA GPRSEAGHLL LQKLLRAKNV QLSTGRGSEG LRAEINGHI Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 90%.

KMT2D is a histone methyltransferase that methylates Lys-4 of histone H3 (H3K4me). H3K4me represents a specific tag for epigenetic transcriptional activation. KMT2D acts as a coactivator for estrogen receptors by being recruited by ESR1 and activating transcription. KMT2D is involved in chromatin organization, chormatin silencig, oocyte growth, oogenesis, and a positive regulation of cell proliferation. Mutations in the KMT2D gene cause Kabuki syndrome (KABUK1) which results in congential mental retardation syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2D
Applications Immunocytochemistry
Classification Monoclonal
Clone CL5346
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KMT2D
Gene Accession No. O14686
Gene Alias AAD10; ALL1-related protein; ALR; CAGL114; Histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; KABUK1; Kabuki make-up syndrome; Kabuki mental retardation syndrome; KMS; KMT2D; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2D; lysine N-methyltransferase 2D; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia 2; Myeloid/lymphoid or mixed-lineage leukemia protein 2; TNRC21; trinucleotide repeat containing 21
Gene Symbols KMT2D
Host Species Mouse
Immunogen Recombinant protein fragment of Human KMT2D (Product #RP-102441)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.