Learn More
Invitrogen™ KMT2D Polyclonal Antibody

Description
KMT2D Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | KMT2D |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | KMT2D |
| Gene Accession No. | O14686 |
| Gene Alias | AAD10; ALL1-related protein; ALR; CAGL114; Histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; KABUK1; Kabuki make-up syndrome; Kabuki mental retardation syndrome; KMS; KMT2D; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2D; lysine N-methyltransferase 2D; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia 2; Myeloid/lymphoid or mixed-lineage leukemia protein 2; TNRC21; trinucleotide repeat containing 21 |
| Gene Symbols | KMT2D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.