Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KMT2E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117037 Supplier Invitrogen™ Supplier No. PA5117037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: NCI-H460, 293T, Mouse brain Immunogen sequence: VELYTAFQHT PTSITLTASR VSKVNDKRRK KSGEKEQHIS KCKKAFREGS RKSSRVKGSA PEIDPSSDGS NFGWETKIKA WMDRYEEANN NQYSEGVQRE AQRIALRLGN GNDKKEMNKS DLNTNNLLFK PPVESHIQKN KKILKSAKDL PPDALIIEYR GKFMLREQFE ANGYFFKRPY PFVLFYSKFH GLEMCVDART FGNEARFIRR SCTPNAEVRH EIQDGTIHLY IYSIHSIPKG TEITIAFDFD YGNCKYKVDC ACLKENPECP VLKRSSESME NINSGYETRR KKGKKDKDIS K.

The mixed lineage leukemia (MLL) gene family comprise a group of Histone H3 lysine 4 (H3K4) methyltransferases within the larger SET1 family. The founding member MLL commonly undergoes translocations in infantile leukemia and displays increased expression in some adult myeloid leukemias. MLL2, also designated ALR, exists within a complex of proteins. MLL2 is important for mouse embryonic development and may be involved in adhesion-related cytoskeletal events affecting cell growth and survival. The MLL2 gene maps to the human locus 19q13.1, which is a frequent target of rearrangement or amplification in solid tumors. MLL3 or its paralogue MLL4 associate with activating signal cointegrator-2 (ASC-2), which regulates ligand-dependent H3K4 trimethylation and expression of LXR-target genes. MLL3 maps to a location on human chromosome 7 that is often deleted in myeloid disorders. MLL3 also exhibits higher expression in peripheral blood, placenta, pancreas, testis, and fetal thymus. MLL5 localizes to the nucleus and forms intranuclear protein complexes, which may regulate chromatin remodeling and cellular growth suppression. The gene encoding human MLL5 lies within chromosome band 7q22.3, a region deleted in cytogenetic aberrations of acute myeloid malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2E
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KMT2E
Gene Accession No. Q3UG20, Q8IZD2
Gene Alias 1810033J14Rik; 9530077A04Rik; D230038D11Rik; HDCMC04P; histone-lysine N-methyltransferase 2E; histone-lysine N-methyltransferase MLL5; Inactive histone-lysine N-methyltransferase 2E; Inactive lysine N-methyltransferase 2E; KMT2E; lysine (K)-specific methyltransferase 2E; lysine methyltransferase 2E; lysine N-methyltransferase 2E; MGC70452; MLL5; myeloid/lymphoid or mixed-lineage leukemia 5; myeloid/lymphoid or mixed-lineage leukemia 5 (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia protein 5; Myeloid/lymphoid or mixed-lineage leukemia protein 5 homolog; NKp44L
Gene Symbols KMT2E
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human MLL5 (NP_0611523).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311968, 55904, 69188
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.